Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATAAAAAGGCCTGACCTTCGGC[C/T]GAAGGATTTACCACATTCTTTACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602444 | ||||||||||||||||||||
Literature Links: |
LOC102724657 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102724657 - uncharacterized LOC102724657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF354A - zinc finger protein 354A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324339.1 | 1627 | Missense Mutation | AGC,GGC | S,G 266 | NP_001311268.1 | |
NM_005649.2 | 1627 | Missense Mutation | AGC,GGC | S,G 308 | NP_005640.2 | |
XM_011534645.2 | 1627 | Missense Mutation | AGC,GGC | S,G 266 | XP_011532947.1 | |
XM_017009791.1 | 1627 | Missense Mutation | AGC,GGC | S,G 308 | XP_016865280.1 |