Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGGGTTTGATGGCGAATATCAA[G/T]ATGACAGCTTAGATTTGCTACGCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DNAJC21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC21 - DnaJ heat shock protein family (Hsp40) member C21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012339.2 | 474 | Missense Mutation | GAT,TAT | D,Y 83 | NP_001012339.2 | |
NM_194283.3 | 474 | Missense Mutation | GAT,TAT | D,Y 83 | NP_919259.3 | |
XM_005248249.4 | 474 | Missense Mutation | GAT,TAT | D,Y 170 | XP_005248306.2 | |
XM_005248250.3 | 474 | Missense Mutation | GAT,TAT | D,Y 170 | XP_005248307.2 | |
XM_011513965.2 | 474 | Missense Mutation | GAT,TAT | D,Y 170 | XP_011512267.2 | |
XM_011513966.2 | 474 | Missense Mutation | GAT,TAT | D,Y 170 | XP_011512268.2 |