Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTTTTTCAATATTTTGTCGTA[C/T]GTTGTCTCAGCTGCATCGCAAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600837 | ||||||||||||||||||||
Literature Links: |
GDNF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDNF - glial cell derived neurotrophic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000514.3 | 685 | Silent Mutation | ACA,ACG | T,T 155 | NP_000505.1 | |
NM_001190468.1 | 685 | Silent Mutation | ACA,ACG | T,T 172 | NP_001177397.1 | |
NM_001190469.1 | 685 | Silent Mutation | ACA,ACG | T,T 146 | NP_001177398.1 | |
NM_001278098.1 | 685 | Silent Mutation | ACA,ACG | T,T 103 | NP_001265027.1 | |
NM_199231.2 | 685 | Silent Mutation | ACA,ACG | T,T 129 | NP_954701.1 | |
XM_011514030.2 | 685 | Silent Mutation | ACA,ACG | T,T 103 | XP_011512332.1 | |
XM_017009337.1 | 685 | Silent Mutation | ACA,ACG | T,T 129 | XP_016864826.1 |