Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCTTCACCTTTCTGGCCCCAC[A/G]CAGCCACCAGACAGCCATGCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616384 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT3A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGT3A2 - UDP glycosyltransferase family 3 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168316.1 | 1675 | Missense Mutation | CGT,TGT | R,C 481 | NP_001161788.1 | |
NM_174914.3 | 1675 | Missense Mutation | CGT,TGT | R,C 515 | NP_777574.2 | |
XM_011513988.2 | 1675 | Missense Mutation | CGT,TGT | R,C 542 | XP_011512290.1 | |
XM_017009150.1 | 1675 | Missense Mutation | CGT,TGT | R,C 477 | XP_016864639.1 |