Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTTACAGCTGGAGACTGAGAAG[C/T]AGGCATCTGTACTGTAAACCTTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603139 MIM: 600822 | ||||||||||||||||||||
Literature Links: |
AK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AK6 - adenylate kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015891.1 | 1044 | Intron | NP_001015891.1 | |||
NM_016283.4 | 1044 | Intron | NP_057367.1 |
RAD17 - RAD17 checkpoint clamp loader component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF9 - TATA-box binding protein associated factor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015892.1 | 1044 | Missense Mutation | ACT,GCT | T,A 193 | NP_001015892.1 | |
NM_003187.4 | 1044 | Intron | NP_003178.1 |