Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGGCGGGTTTGCACACGTGGT[A/C]TCCTCCGAGCCTTCAAGTTTGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602226 | ||||||||||||||||||||
Literature Links: |
CD180 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD180 - CD180 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005582.2 | 1969 | Silent Mutation | NP_005573.2 | |||
XM_005248504.4 | 1969 | Missense Mutation | XP_005248561.1 |