Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCACTGGTGATAAACGAGCCCC[A/G]TTCCTTGTTGTGATCATGATGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604489 MIM: 606202 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMACR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMACR - alpha-methylacyl-CoA racemase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167595.1 | 1030 | Missense Mutation | CGG,TGG | R,W 312 | NP_001161067.1 | |
NM_014324.5 | 1030 | Missense Mutation | CGG,TGG | R,W 312 | NP_055139.4 | |
NM_203382.2 | 1030 | UTR 3 | NP_976316.1 |
C1QTNF3-AMACR - C1QTNF3-AMACR readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC45A2 - solute carrier family 45 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |