Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCCGAAGCGCGTGTTACTGTT[C/T]CCTGCTGTCCAGGCAAGATTGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf15 - chromosome 5 open reading frame 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105379183 - uncharacterized LOC105379183 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VDAC1 - voltage dependent anion channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003374.2 | 884 | Silent Mutation | GGA,GGG | G,G 213 | NP_003365.1 | |
XM_005272075.3 | 884 | Intron | XP_005272132.1 | |||
XM_017009821.1 | 884 | Intron | XP_016865310.1 | |||
XM_017009822.1 | 884 | Intron | XP_016865311.1 | |||
XM_017009823.1 | 884 | Intron | XP_016865312.1 |