Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCCCTCGGTTCCCAGCTCAGG[G/T]TCCCCACGACCCCTGGTGTGGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf47 - chromosome 5 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144954.1 | 328 | Missense Mutation | GTC,TTC | V,F 77 | NP_001138426.1 | |
XM_011534431.1 | 328 | Missense Mutation | GTC,TTC | V,F 77 | XP_011532733.1 | |
XM_017009028.1 | 328 | Missense Mutation | GTC,TTC | V,F 77 | XP_016864517.1 |