Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGTGGTGGTGCCTCCTGGAGA[A/G]GAACAGAGATACACATGCCATGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLA-F - major histocompatibility complex, class I, F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098478.1 | 852 | Intron | NP_001091948.1 | |||
NM_001098479.1 | 852 | Silent Mutation | GAA,GAG | E,E 274 | NP_001091949.1 | |
NM_018950.2 | 852 | Silent Mutation | GAA,GAG | E,E 274 | NP_061823.2 | |
XM_011514564.1 | 852 | Silent Mutation | GAA,GAG | E,E 274 | XP_011512866.1 | |
XM_017010810.1 | 852 | Silent Mutation | GAA,GAG | E,E 274 | XP_016866299.1 | |
XM_017010811.1 | 852 | Silent Mutation | GAA,GAG | E,E 274 | XP_016866300.1 | |
XM_017010812.1 | 852 | Silent Mutation | GAA,GAG | E,E 274 | XP_016866301.1 | |
XM_017010813.1 | 852 | Silent Mutation | GAA,GAG | E,E 274 | XP_016866302.1 | |
XM_017010814.1 | 852 | Silent Mutation | GAA,GAG | E,E 156 | XP_016866303.1 | |
XM_017010815.1 | 852 | Silent Mutation | GAA,GAG | E,E 156 | XP_016866304.1 |
HLA-F-AS1 - HLA-F antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |