Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCAGCACCCCTCCAACTGTGC[C/G]CCACCAGGGGCTGCCCGCAGCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603382 MIM: 609693 | ||||||||||||||||||||
Literature Links: |
MSH5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSH5 - mutS homolog 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSH5-SAPCD1 - MSH5-SAPCD1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAPCD1 - suppressor APC domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039651.1 | 2489 | Intron | NP_001034740.1 |
SAPCD1-AS1 - SAPCD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA7 - von Willebrand factor A domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025258.2 | 2489 | Missense Mutation | CGC,GGC | R,G 877 | NP_079534.2 | |
XM_005249427.2 | 2489 | Missense Mutation | CGC,GGC | R,G 903 | XP_005249484.1 | |
XM_017011327.1 | 2489 | Missense Mutation | CGC,GGC | R,G 903 | XP_016866816.1 | |
XM_017011328.1 | 2489 | Missense Mutation | CGC,GGC | R,G 903 | XP_016866817.1 | |
XM_017011329.1 | 2489 | Missense Mutation | CGC,GGC | R,G 629 | XP_016866818.1 |