Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATGAGAGTTGGATCTGGCATC[C/G]TGACAGAGGGTTCCAGTGATGGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159465 MIM: 612192 | ||||||||||||||||||||
Literature Links: |
MOG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MOG - myelin oligodendrocyte glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008228.2 | 1067 | Intron | NP_001008229.1 | |||
NM_001008229.2 | 1067 | Intron | NP_001008230.1 | |||
NM_001170418.1 | 1067 | Intron | NP_001163889.1 | |||
NM_002433.4 | 1067 | Intron | NP_002424.3 | |||
NM_206809.3 | 1067 | Intron | NP_996532.2 | |||
NM_206810.3 | 1067 | Intron | NP_996533.2 | |||
NM_206811.3 | 1067 | Intron | NP_996534.2 | |||
NM_206812.3 | 1067 | Intron | NP_996535.2 | |||
NM_206814.5 | 1067 | Intron | NP_996537.3 | |||
XM_005249131.3 | 1067 | Intron | XP_005249188.1 |
ZFP57 - ZFP57 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109809.2 | 1067 | Missense Mutation | CAC,CAG | H,Q 367 | NP_001103279.2 | |
XM_006715087.3 | 1067 | Missense Mutation | CAC,CAG | H,Q 295 | XP_006715150.1 | |
XM_011514570.2 | 1067 | Missense Mutation | CAC,CAG | H,Q 367 | XP_011512872.1 |