Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACTCCAGCATGTCCTCCTACTC[G/T]CTGGAGCAGAGCTACTTGCACCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXF2 - forkhead box F2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001452.1 | 1239 | Silent Mutation | TCG,TCT | S,S 375 | NP_001443.1 |
LOC105374880 - uncharacterized LOC105374880 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6720 - microRNA 6720 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |