Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTGGCAGTGAATATTGCTGAT[G/T]AGGATGAGAGGCGTAGGTGAAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCM1 - glial cells missing homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003643.3 | 1401 | Missense Mutation | AAT,CAT | N,H 398 | NP_003634.2 | |
XM_017011390.1 | 1401 | Missense Mutation | AAT,CAT | N,H 460 | XP_016866879.1 |