Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGGAATGCACACAACTTCCCAT[A/G]GTGCATCTACTGCAGTGAGTGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUC21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUC21 - mucin 21, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010909.3 | 1698 | Missense Mutation | AGT,GGT | S,G 461 | NP_001010909.2 | |
NM_001322370.1 | 1698 | Missense Mutation | NP_001309299.1 | |||
NM_001322371.1 | 1698 | Missense Mutation | NP_001309300.1 |