Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTCCGAGAAGGCATCGACAAGC[A/C]GGACCCTCCCACCTGGAAGACGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601900 | ||||||||||||||||||||
Literature Links: |
IRF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IRF4 - interferon regulatory factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195286.1 | 389 | Missense Mutation | CAG,CCG | Q,P 88 | NP_001182215.1 | |
NM_002460.3 | 389 | Missense Mutation | CAG,CCG | Q,P 88 | NP_002451.2 | |
XM_006715090.2 | 389 | Missense Mutation | CAG,CCG | Q,P 88 | XP_006715153.1 |