Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTCATCTACTTTCTGCTTTTT[A/C]TCTTTCTTTTCTTTGGTATCTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608994 MIM: 600772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKS1A - ankyrin repeat and sterile alpha motif domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105375030 - uncharacterized LOC105375030 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF11 - TATA-box binding protein associated factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270488.1 | 783 | Missense Mutation | GAG,GAT | E,D 93 | NP_001257417.1 | |
NM_005643.3 | 783 | Missense Mutation | GAG,GAT | E,D 93 | NP_005634.1 | |
XM_011514827.2 | 783 | Missense Mutation | GAG,GAT | E,D 49 | XP_011513129.1 |
UHRF1BP1 - UHRF1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |