Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTCCAGATGTTCGAGACCTTCC[A/G]AGGAGGACTGTAGCCGCCCGGCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612808 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRFN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRFN2 - leucine rich repeat and fibronectin type III domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020737.2 | 3647 | Missense Mutation | CGG,TGG | R,W 746 | NP_065788.1 | |
XM_011514761.2 | 3647 | Missense Mutation | CGG,TGG | R,W 746 | XP_011513063.1 | |
XM_011514762.2 | 3647 | Missense Mutation | CGG,TGG | R,W 746 | XP_011513064.1 | |
XM_017011110.1 | 3647 | Missense Mutation | CGG,TGG | R,W 746 | XP_016866599.1 | |
XM_017011111.1 | 3647 | Intron | XP_016866600.1 |