Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTGCCGAGAAGACTCCCGTGAA[A/G]AAGAAGGCCCGCAAGTCTGCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142220 MIM: 602799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H1E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H1E - histone cluster 1, H1e | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005321.2 | 123 | Silent Mutation | AAA,AAG | K,K 21 | NP_005312.1 |
HIST1H2BD - histone cluster 1, H2bd | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |