Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGGGCTGAGTAGGAGCACAGA[C/T]GTGCTGGGGGCCTGCTCACACAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612035 MIM: 186975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AARS2 - alanyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020745.3 | 2315 | Silent Mutation | ACA,ACG | T,T 910 | NP_065796.1 | |
XM_005249245.3 | 2315 | Silent Mutation | ACA,ACG | T,T 813 | XP_005249302.1 | |
XM_011514764.2 | 2315 | Silent Mutation | ACA,ACG | T,T 910 | XP_011513066.1 | |
XM_017011112.1 | 2315 | Silent Mutation | ACA,ACG | T,T 480 | XP_016866601.1 |
TCTE1 - t-complex-associated-testis-expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |