Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCGTCCTGGACGTGCTGCTGCT[C/G]CATCGCCTGGTCCTCCTGGAGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611200 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101926934 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101926934 - uncharacterized LOC101926934 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDRD6 - tudor domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010870.2 | 782 | Silent Mutation | CTC,CTG | L,L 176 | NP_001010870.1 | |
NM_001168359.1 | 782 | Silent Mutation | CTC,CTG | L,L 176 | NP_001161831.1 |