Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGATGGCAGGGCCCCAGGCAGT[A/C]CAGGACCCCAGGCTCTGAAGGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611419 MIM: 600701 | ||||||||||||||||||||
Literature Links: |
C6orf1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf1 - chromosome 6 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008703.2 | 805 | Missense Mutation | NP_001008703.1 | |||
NM_001008704.2 | 805 | Missense Mutation | NP_001008704.1 | |||
NM_001287396.1 | 805 | Missense Mutation | NP_001274325.1 | |||
NM_001287397.1 | 805 | Missense Mutation | NP_001274326.1 | |||
NM_001287398.1 | 805 | Missense Mutation | NP_001274327.1 | |||
NM_178508.4 | 805 | Missense Mutation | NP_848603.2 |
HMGA1 - high mobility group AT-hook 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319077.1 | 805 | Intron | NP_001306006.1 | |||
NM_001319078.1 | 805 | Intron | NP_001306007.1 | |||
NM_001319079.1 | 805 | Intron | NP_001306008.1 | |||
NM_001319080.1 | 805 | Intron | NP_001306009.1 | |||
NM_001319081.1 | 805 | Intron | NP_001306010.1 | |||
NM_001319082.1 | 805 | Intron | NP_001306011.1 | |||
NM_002131.3 | 805 | Intron | NP_002122.1 | |||
NM_145899.2 | 805 | Intron | NP_665906.1 | |||
NM_145901.2 | 805 | Intron | NP_665908.1 | |||
NM_145902.2 | 805 | Intron | NP_665909.1 | |||
NM_145903.2 | 805 | Intron | NP_665910.1 | |||
NM_145905.2 | 805 | Intron | NP_665912.1 |
MIR6835 - microRNA 6835 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |