Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGGTTTTAGCAGTCTTTGGTGA[C/G]TTGGAGTCCCTGGATAAAACCAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142712 MIM: 602827 | ||||||||||||||||||||
Literature Links: |
HIST1H1T PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST1H1T - histone cluster 1, H1t | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005323.3 | 469 | Missense Mutation | AAC,AAG | N,K 142 | NP_005314.2 |
HIST1H4C - histone cluster 1, H4c | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |