Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGACCTGGGCTCCACTGTCAT[A/C]AAAGAAGTCTTGAAGAGGGCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100678 MIM: 147460 MIM: 605442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACAT2 - acetyl-CoA acetyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303253.1 | 245 | Silent Mutation | ATA,ATC | I,I 67 | NP_001290182.1 | |
NM_005891.2 | 245 | Silent Mutation | ATA,ATC | I,I 38 | NP_005882.2 |
LOC100129518 - uncharacterized LOC100129518 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOD2 - superoxide dismutase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000636.3 | 245 | Intron | NP_000627.2 | |||
NM_001024465.2 | 245 | Intron | NP_001019636.1 | |||
NM_001024466.2 | 245 | Intron | NP_001019637.1 | |||
NM_001322814.1 | 245 | Intron | NP_001309743.1 | |||
NM_001322815.1 | 245 | Intron | NP_001309744.1 | |||
NM_001322816.1 | 245 | Intron | NP_001309745.1 | |||
NM_001322817.1 | 245 | Intron | NP_001309746.1 | |||
NM_001322819.1 | 245 | Intron | NP_001309748.1 | |||
NM_001322820.1 | 245 | Intron | NP_001309749.1 |
WTAP - Wilms tumor 1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |