Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGTGTTGGGTGCGGGGCCTGGG[C/T]CGCCGCTCTCGCTCCTCTGGCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LACE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LACE1 - lactation elevated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323005.1 | 178 | Missense Mutation | GCC,GTC | A,V 30 | NP_001309934.1 | |
NM_145315.4 | 178 | Missense Mutation | GCC,GTC | A,V 30 | NP_660358.2 | |
XM_005266885.3 | 178 | Intron | XP_005266942.1 | |||
XM_011535657.2 | 178 | Missense Mutation | GCC,GTC | A,V 30 | XP_011533959.1 | |
XM_011535659.2 | 178 | Missense Mutation | GCC,GTC | A,V 30 | XP_011533961.1 | |
XM_011535660.2 | 178 | Missense Mutation | GCC,GTC | A,V 30 | XP_011533962.1 | |
XM_011535661.2 | 178 | Missense Mutation | GCC,GTC | A,V 30 | XP_011533963.1 | |
XM_017010647.1 | 178 | Intron | XP_016866136.1 | |||
XM_017010648.1 | 178 | Intron | XP_016866137.1 | |||
XM_017010649.1 | 178 | UTR 5 | XP_016866138.1 |