Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCAGCAGTTCGGCATTTGGGATC[A/G]AAAGCAGAAAAGCATGTCCATGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1586 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1586 - KIAA1586 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286274.1 | 718 | Silent Mutation | TCA,TCG | S,S 145 | NP_001273203.1 | |
NM_001286275.1 | 718 | Silent Mutation | TCA,TCG | S,S 105 | NP_001273204.1 | |
NM_001286276.1 | 718 | Silent Mutation | TCA,TCG | S,S 105 | NP_001273205.1 | |
NM_020931.3 | 718 | Silent Mutation | TCA,TCG | S,S 172 | NP_065982.1 | |
XM_011514767.2 | 718 | Intron | XP_011513069.1 |