Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGAGTAGCAGACATTGTCCCTC[A/G]GAAGGGGTGACCCCACGGGCATGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611981 MIM: 612648 | ||||||||||||||||||||
Literature Links: |
MRPS18A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPS18A - mitochondrial ribosomal protein S18A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193343.1 | 575 | UTR 3 | NP_001180272.1 | |||
NM_018135.3 | 575 | Missense Mutation | CCG,CTG | P,L 181 | NP_060605.1 | |
XM_006715134.2 | 575 | Missense Mutation | CCG,CTG | P,L 255 | XP_006715197.1 |
RSPH9 - radial spoke head 9 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193341.1 | 575 | Intron | NP_001180270.1 | |||
NM_152732.4 | 575 | Intron | NP_689945.2 | |||
XM_005248901.3 | 575 | Intron | XP_005248958.1 | |||
XM_006715014.1 | 575 | Intron | XP_006715077.1 | |||
XM_017010426.1 | 575 | Intron | XP_016865915.1 |