Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGCTCCAGGGAGTGGGCCGGA[C/G]TCTGTTTGCCCCGCTGGGACTCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611687 MIM: 611689 | ||||||||||||||||||||
Literature Links: |
KHDC3L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KHDC3L - KH domain containing 3 like, subcortical maternal complex member | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OOEP - oocyte expressed protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080507.2 | 143 | Missense Mutation | ACT,AGT | T,S 16 | NP_001073976.1 | |
XM_017010890.1 | 143 | Missense Mutation | ACT,AGT | T,S 16 | XP_016866379.1 |
OOEP-AS1 - OOEP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |