Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACAAAACCATGGAAGTGACTGG[G/T]TTTGGAGTGACCAGGTGTTCCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609577 MIM: 611822 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL7 - cullin 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLC4 - kinesin light chain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289034.1 | 368 | Intron | NP_001275963.1 | |||
NM_001289035.1 | 368 | Intron | NP_001275964.1 | |||
NM_138343.3 | 368 | Intron | NP_612352.1 | |||
NM_201521.2 | 368 | Intron | NP_958929.1 | |||
NM_201522.2 | 368 | UTR 5 | NP_958930.1 | |||
NM_201523.2 | 368 | Silent Mutation | GGG,GGT | G,G 5 | NP_958931.1 | |
XM_011514989.1 | 368 | Silent Mutation | GGG,GGT | G,G 5 | XP_011513291.1 | |
XM_017011457.1 | 368 | Intron | XP_016866946.1 |
MRPL2 - mitochondrial ribosomal protein L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300848.1 | 368 | Intron | NP_001287777.1 | |||
NM_015950.4 | 368 | Intron | NP_057034.2 | |||
XM_005249161.4 | 368 | Intron | XP_005249218.1 |