Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTGTTGGGAACTCTTTAGGCCT[C/T]GGTTTCTGGTGACAGCCCCAGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608859 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD109 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD109 - CD109 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159587.2 | 357 | Missense Mutation | CGG,TGG | R,W 27 | NP_001153059.1 | |
NM_001159588.2 | 357 | Missense Mutation | CGG,TGG | R,W 27 | NP_001153060.1 | |
NM_133493.4 | 357 | Missense Mutation | CGG,TGG | R,W 27 | NP_598000.2 | |
XM_005248659.3 | 357 | Missense Mutation | CGG,TGG | R,W 27 | XP_005248716.1 | |
XM_011535473.1 | 357 | Missense Mutation | CGG,TGG | R,W 14 | XP_011533775.1 | |
XM_017010283.1 | 357 | Intron | XP_016865772.1 |
LOC101928489 - uncharacterized LOC101928489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |