Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCCATAGCTGGCAGGGCCTGGG[G/T]CACCTGAAACACACAGAAGATTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120165 MIM: 120210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL19A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL19A1 - collagen type XIX alpha 1 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COL9A1 - collagen type IX alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001851.4 | 2131 | Missense Mutation | GAC,GCC | D,A 862 | NP_001842.3 | |
NM_078485.3 | 2131 | Missense Mutation | GAC,GCC | D,A 619 | NP_511040.2 | |
XM_011535429.2 | 2131 | Missense Mutation | GAC,GCC | D,A 872 | XP_011533731.1 | |
XM_011535430.2 | 2131 | Missense Mutation | GAC,GCC | D,A 629 | XP_011533732.1 | |
XM_017010246.1 | 2131 | Missense Mutation | GAC,GCC | D,A 689 | XP_016865735.1 | |
XM_017010247.1 | 2131 | Missense Mutation | GAC,GCC | D,A 445 | XP_016865736.1 |