Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAAGTCTGGCTACACAGGGGAC[A/G]GCAAACACTGCAAAGGTGAGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614708 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCUBE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCUBE3 - signal peptide, CUB domain and EGF like domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303136.1 | 565 | Missense Mutation | AGC,GGC | S,G 65 | NP_001290065.1 | |
NM_152753.3 | 565 | Missense Mutation | AGC,GGC | S,G 65 | NP_689966.2 | |
XM_005248943.1 | 565 | Missense Mutation | AGC,GGC | S,G 65 | XP_005249000.1 | |
XM_005248947.1 | 565 | Missense Mutation | AGC,GGC | S,G 65 | XP_005249004.1 | |
XM_011514404.2 | 565 | Missense Mutation | AGC,GGC | S,G 57 | XP_011512706.1 |