Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCTTATTCCTCTGCACGGACTC[C/G]GAGACGGTCCTCAGCGGGAGGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAGAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAGAP - T-cell activation RhoGTPase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278733.1 | 2426 | Silent Mutation | TCC,TCG | S,S 635 | NP_001265662.1 | |
NM_054114.4 | 2426 | Silent Mutation | TCC,TCG | S,S 698 | NP_473455.2 | |
NM_138810.3 | 2426 | Intron | NP_620165.1 | |||
NM_152133.2 | 2426 | Silent Mutation | TCC,TCG | S,S 520 | NP_687034.1 |