Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGGATCTCAGAAGGTTCCACTT[C/G]GTAAAAGTCTGTGCTCTCATCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611470 | ||||||||||||||||||||
Literature Links: |
LGSN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGSN - lengsin, lens protein with glutamine synthetase domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143940.1 | 2496 | UTR 3 | NP_001137412.1 | |||
NM_016571.2 | 2496 | Missense Mutation | CAA,GAA | Q,E 449 | NP_057655.2 | |
XM_011535889.2 | 2496 | Missense Mutation | CAA,GAA | Q,E 530 | XP_011534191.1 | |
XM_011535892.2 | 2496 | Missense Mutation | CAA,GAA | Q,E 488 | XP_011534194.1 | |
XM_017010929.1 | 2496 | Missense Mutation | CAA,GAA | Q,E 530 | XP_016866418.1 | |
XM_017010930.1 | 2496 | Missense Mutation | CAA,GAA | Q,E 530 | XP_016866419.1 | |
XM_017010931.1 | 2496 | Missense Mutation | CAA,GAA | Q,E 425 | XP_016866420.1 |