Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCACGCCAAGCGCAAAACTGTCA[C/T]AGCCATGGATGTAGTATATGCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613609 MIM: 142712 MIM: 602827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HFE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HFE - hemochromatosis | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H1T - histone cluster 1, H1t | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4C - histone cluster 1, H4c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003542.3 | 248 | Missense Mutation | ACA,ATA | T,I 83 | NP_003533.1 |