Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600214 MIM: 603099 MIM: 176311 MIM: 602677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGER - advanced glycosylation end product-specific receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136.4 | 1337 | Missense Mutation | CGC,GGC | R,G 397 | NP_001127.1 | |
NM_001206929.1 | 1337 | Missense Mutation | CGC,GGC | R,G 413 | NP_001193858.1 | |
NM_001206932.1 | 1337 | Missense Mutation | CGC,GGC | R,G 383 | NP_001193861.1 | |
NM_001206934.1 | 1337 | UTR 3 | NP_001193863.1 | |||
NM_001206936.1 | 1337 | UTR 3 | NP_001193865.1 | |||
NM_001206940.1 | 1337 | UTR 3 | NP_001193869.1 | |||
NM_001206954.1 | 1337 | UTR 3 | NP_001193883.1 | |||
NM_001206966.1 | 1337 | Intron | NP_001193895.1 | |||
NM_172197.2 | 1337 | UTR 3 | NP_751947.1 | |||
XM_017010328.1 | 1337 | UTR 3 | XP_016865817.1 |
AGPAT1 - 1-acylglycerol-3-phosphate O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPSM3 - G-protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6833 - microRNA 6833 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PBX2 - PBX homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF5 - ring finger protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006913.3 | 1337 | Intron | NP_008844.1 |