Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTTCCAGGAACTACCAAAGCCA[C/T]GGATCAGCTGAGTTACCCGGGCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603443 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCG25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCG25 - HLA complex group 25 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS52 - VPS52, GARP complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289174.1 | 2097 | Missense Mutation | CAT,CGT | H,R 576 | NP_001276103.1 | |
NM_001289175.1 | 2097 | Missense Mutation | CAT,CGT | H,R 518 | NP_001276104.1 | |
NM_001289176.1 | 2097 | Missense Mutation | CAT,CGT | H,R 454 | NP_001276105.1 | |
NM_022553.5 | 2097 | Missense Mutation | CAT,CGT | H,R 643 | NP_072047.4 | |
XM_011514797.1 | 2097 | Missense Mutation | CAT,CGT | H,R 576 | XP_011513099.1 | |
XM_011514798.2 | 2097 | Missense Mutation | CAT,CGT | H,R 576 | XP_011513100.1 | |
XM_011514799.1 | 2097 | Missense Mutation | CAT,CGT | H,R 576 | XP_011513101.1 | |
XM_017011177.1 | 2097 | Intron | XP_016866666.1 | |||
XM_017011178.1 | 2097 | Missense Mutation | CAT,CGT | H,R 518 | XP_016866667.1 | |
XM_017011179.1 | 2097 | Intron | XP_016866668.1 |