Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCCCCTCACTTCATGTTCTCGA[C/T]GTTGCGTCCTCGCACGGCACACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600214 MIM: 603099 MIM: 609897 MIM: 603298 MIM: 602677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGER - advanced glycosylation end product-specific receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AGPAT1 - 1-acylglycerol-3-phosphate O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006411.3 | 825 | Missense Mutation | ATC,GTC | I,V 63 | NP_006402.1 | |
NM_032741.4 | 825 | Missense Mutation | ATC,GTC | I,V 63 | NP_116130.2 | |
XM_005248805.3 | 825 | Missense Mutation | ATC,GTC | I,V 67 | XP_005248862.1 | |
XM_005248806.2 | 825 | Missense Mutation | ATC,GTC | I,V 63 | XP_005248863.1 | |
XM_011514234.1 | 825 | Missense Mutation | ATC,GTC | I,V 63 | XP_011512536.1 |
EGFL8 - EGF like domain multiple 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6721 - microRNA 6721 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6833 - microRNA 6833 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPT2 - palmitoyl-protein thioesterase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPT2-EGFL8 - PPT2-EGFL8 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF5 - ring finger protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |