Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTTGGAAGCACTGTCTTTTTAG[C/T]GCGTCTCTTATCTCTGCCCTGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCNT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCNT2 - glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001491.2 | 582 | Intron | NP_001482.1 | |||
NM_145649.4 | 582 | Silent Mutation | AGC,AGT | S,S 11 | NP_663624.1 | |
NM_145655.3 | 582 | Intron | NP_663630.2 | |||
XM_005248997.3 | 582 | Intron | XP_005249054.1 | |||
XM_005248999.2 | 582 | Intron | XP_005249056.1 | |||
XM_006715052.3 | 582 | Silent Mutation | AGC,AGT | S,S 11 | XP_006715115.1 | |
XM_011514466.2 | 582 | Intron | XP_011512768.1 | |||
XM_011514468.2 | 582 | Silent Mutation | AGC,AGT | S,S 11 | XP_011512770.1 | |
XM_017010732.1 | 582 | Silent Mutation | AGC,AGT | S,S 11 | XP_016866221.1 |