Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCATCTGTAAGCCTTTCAGCAC[A/G]CTGGTGAAAAGCCACGTAACTGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610060 MIM: 607845 MIM: 609775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR1C - RNA polymerase I subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318876.1 | 3436 | Intron | NP_001305805.1 | |||
NM_203290.3 | 3436 | Intron | NP_976035.1 | |||
XM_011515000.2 | 3436 | Intron | XP_011513302.1 |
XPO5 - exportin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020750.2 | 3436 | Silent Mutation | AGC,AGT | S,S 1075 | NP_065801.1 |
YIPF3 - Yip1 domain family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |