Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGCCGTGGACTTCCTTAGCAACC[C/T]TCAGGGCAAGGAGGTGCACGGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611200 | ||||||||||||||||||||
Literature Links: |
LOC101926934 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101926934 - uncharacterized LOC101926934 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDRD6 - tudor domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010870.2 | 736 | Missense Mutation | CCT,CTT | P,L 161 | NP_001010870.1 | |
NM_001168359.1 | 736 | Missense Mutation | CCT,CTT | P,L 161 | NP_001161831.1 |