Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAGCATCCTCCTCTTGACCCA[C/T]GGCTCACCAAAAAGTAAGTCAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D22B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBC1D22B - TBC1 domain family member 22B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017772.3 | 314 | Missense Mutation | CGG,TGG | R,W 34 | NP_060242.2 | |
XM_011514738.2 | 314 | Missense Mutation | CGG,TGG | R,W 34 | XP_011513040.1 | |
XM_011514739.1 | 314 | Missense Mutation | CGG,TGG | R,W 34 | XP_011513041.1 |