Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTTTTTGGCTGAGGTGTTTTC[A/C]CCTTTTTCGCACTCTTGGCCACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142210 MIM: 602813 MIM: 602824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H1D - histone cluster 1, H1d | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005320.2 | 588 | Missense Mutation | GGG,GTG | G,V 178 | NP_005311.1 |
HIST1H3E - histone cluster 1, H3e | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4F - histone cluster 1, H4f | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |