Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTAGGGAGGATAGGGAGAAGAT[A/G]TGCCTCGCTATTGAAGATCCAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604344 | ||||||||||||||||||||
Literature Links: |
MAN1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAN1A1 - mannosidase alpha class 1A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005907.3 | 2209 | Missense Mutation | CAT,TAT | H,Y 636 | NP_005898.2 | |
XM_005266986.4 | 2209 | Missense Mutation | CAT,TAT | H,Y 719 | XP_005267043.1 | |
XM_011535833.2 | 2209 | Missense Mutation | CAT,TAT | H,Y 447 | XP_011534135.1 | |
XM_011535834.2 | 2209 | Intron | XP_011534136.1 |