Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGTGGCTGCTGCCTCATGGACT[G/T]GGACTCCGGTGGAGCCTTCTGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLPSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLPSL1 - colipase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLPSL2 - colipase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286550.1 | 171 | Missense Mutation | TGG,TTG | W,L 49 | NP_001273479.1 | |
NM_207409.3 | 171 | Missense Mutation | TGG,TTG | W,L 49 | NP_997292.2 |