Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTCCTGAGCCAAACCCTTTCCC[A/G]TGATGCATGACCACTGCGGAGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOMM6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOMM6 - translocase of outer mitochondrial membrane 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP49 - ubiquitin specific peptidase 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286554.1 | 1923 | Silent Mutation | CAC,CAT | H,H 607 | NP_001273483.1 | |
NM_018561.4 | 1923 | Silent Mutation | CAC,CAT | H,H 607 | NP_061031.2 |