Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTCCCAAAATAATGGTAGATG[A/G]AACCAATTTGGAATCTGATGTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616823 MIM: 172100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOPEY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOPEY1 - dopey family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199942.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1999 | NP_001186871.1 | |
NM_015018.3 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | NP_055833.2 | |
XM_011535619.2 | 6251 | Missense Mutation | GAA,GGA | E,G 1998 | XP_011533921.1 | |
XM_017010559.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866048.1 | |
XM_017010560.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866049.1 | |
XM_017010561.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1999 | XP_016866050.1 | |
XM_017010562.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1999 | XP_016866051.1 | |
XM_017010563.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866052.1 | |
XM_017010564.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866053.1 | |
XM_017010565.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1999 | XP_016866054.1 | |
XM_017010566.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1999 | XP_016866055.1 | |
XM_017010567.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1999 | XP_016866056.1 | |
XM_017010568.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1998 | XP_016866057.1 | |
XM_017010569.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1998 | XP_016866058.1 | |
XM_017010570.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1975 | XP_016866059.1 | |
XM_017010571.1 | 6251 | Missense Mutation | GAA,GGA | E,G 1973 | XP_016866060.1 | |
XM_017010572.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866061.1 | |
XM_017010573.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866062.1 | |
XM_017010574.1 | 6251 | Missense Mutation | GAA,GGA | E,G 2008 | XP_016866063.1 |
PGM3 - phosphoglucomutase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199917.1 | 6251 | Intron | NP_001186846.1 | |||
NM_001199918.1 | 6251 | Intron | NP_001186847.1 | |||
NM_001199919.1 | 6251 | Intron | NP_001186848.1 | |||
NM_015599.2 | 6251 | Intron | NP_056414.1 | |||
XM_017010935.1 | 6251 | Intron | XP_016866424.1 | |||
XM_017010936.1 | 6251 | Intron | XP_016866425.1 | |||
XM_017010937.1 | 6251 | Intron | XP_016866426.1 | |||
XM_017010938.1 | 6251 | Intron | XP_016866427.1 |