Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGTCTCACAGGCTCACTTACC[A/G]TCTCCATCTGCTGGGGGAGCACTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610060 MIM: 607845 MIM: 609775 | ||||||||||||||||||||
Literature Links: |
POLR1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR1C - RNA polymerase I subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318876.1 | 3193 | Intron | NP_001305805.1 | |||
NM_203290.3 | 3193 | Intron | NP_976035.1 | |||
XM_011515000.2 | 3193 | Intron | XP_011513302.1 |
XPO5 - exportin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020750.2 | 3193 | Silent Mutation | GAC,GAT | D,D 994 | NP_065801.1 |
YIPF3 - Yip1 domain family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |