Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTTCTCACATAAATTTCATCAGG[C/G]GGAGGCTTCTTCTTGCTGCCAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTA4 - glutathione S-transferase alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001512.3 | 771 | Silent Mutation | CCC,CCG | P,P 207 | NP_001503.1 | |
XM_005249035.3 | 771 | Silent Mutation | CCC,CCG | P,P 207 | XP_005249092.1 | |
XM_011514534.2 | 771 | Silent Mutation | CCC,CCG | P,P 170 | XP_011512836.1 | |
XM_011514535.2 | 771 | Silent Mutation | CCC,CCG | P,P 170 | XP_011512837.1 |