Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGATGTGGTTGCAGGACAGGTTGA[C/T]GCTGGTGAGTGTGGTGTTGATGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCTE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCTE1 - t-complex-associated-testis-expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182539.3 | 1875 | Missense Mutation | ATC,GTC | I,V 423 | NP_872345.2 | |
XM_005248872.1 | 1875 | Missense Mutation | ATC,GTC | I,V 423 | XP_005248929.1 | |
XM_005248873.2 | 1875 | Intron | XP_005248930.1 | |||
XM_005248874.2 | 1875 | Missense Mutation | ATC,GTC | I,V 324 | XP_005248931.1 | |
XM_006715009.3 | 1875 | Missense Mutation | ATC,GTC | I,V 324 | XP_006715072.1 | |
XM_011514336.2 | 1875 | Missense Mutation | ATC,GTC | I,V 423 | XP_011512638.1 | |
XM_011514338.2 | 1875 | Missense Mutation | ATC,GTC | I,V 423 | XP_011512640.1 | |
XM_011514339.2 | 1875 | Missense Mutation | ATC,GTC | I,V 324 | XP_011512641.1 | |
XM_011514340.1 | 1875 | Intron | XP_011512642.1 |
TMEM151B - transmembrane protein 151B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |